Assay Details
| Gene Symbol: LOC4339821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783930.2 | XP_015639416.1 | 2 - 3 | 519 | 143 | |
| GenBank mRNA | AK107823.1 | - | 2 - 3 | 464 | 143 | |
| AK121149.1 | - | 2 - 3 | 465 | 143 | ||
More Information
Set Membership: |
|

LOC4339821
Genomic Map