Assay Details
| Gene Symbol: LOC4340052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787395.2 | XP_015642881.1 | 2 - 3 | 529 | 69 | |
| GenBank mRNA | AK069328.1 | - | 2 - 3 | 441 | 69 | |
More Information
Set Membership: |
|

LOC4340052
Genomic Map