Assay Details
| Gene Symbol: LOC4340833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786804.1 | XP_015642290.1 | 14 - 15 | 1550 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4340833| Gene Symbol: LOC4340833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786804.1 | XP_015642290.1 | 14 - 15 | 1550 | 71 | |
Set Membership: |
|