Assay Details
| Gene Symbol: LOC4340904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787163.2 | XP_015642649.1 | 2 | 1653 | 63 | |
| GenBank mRNA | AK109109.1 | - | 1 | 1050 | 63 | |
More Information
Set Membership: |
|

LOC4340904
Genomic Map