Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788862.2 | XP_015644348.1 | 11 - 12 | 1978 | 137 | |
| XM_015788863.2 | XP_015644349.1 | 11 - 12 | 1954 | 137 | ||
| XM_015788864.2 | XP_015644350.1 | 11 - 12 | 1978 | 137 | ||
| GenBank mRNA | AK066930.1 | - | 11 - 12 | 1955 | 137 | |
More Information
Set Membership: |
|

LOC4341008
Genomic Map