Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788862.2 | XP_015644348.1 | 17 - 18 | 2553 | 97 | |
| XM_015788863.2 | XP_015644349.1 | 17 - 18 | 2529 | 97 | ||
| GenBank mRNA | AK061965.1 | - | 4 - 5 | 315 | 97 | |
| AK066930.1 | - | 17 - 18 | 2530 | 97 | ||
More Information
Set Membership: |
|

LOC4341008
Genomic Map