Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788862.2 | XP_015644348.1 | 18 - 19 | 2650 | 84 | |
| XM_015788863.2 | XP_015644349.1 | 18 - 19 | 2626 | 84 | ||
| GenBank mRNA | AK061965.1 | - | 5 - 6 | 412 | 84 | |
| AK066930.1 | - | 18 - 19 | 2627 | 84 | ||
More Information
Set Membership: |
|

LOC4341008
Genomic Map