Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788862.2 | XP_015644348.1 | 20 - 21 | 2797 | 108 | |
| XM_015788863.2 | XP_015644349.1 | 20 - 21 | 2773 | 108 | ||
| XM_015788864.2 | XP_015644350.1 | 19 - 20 | 2692 | 108 | ||
| GenBank mRNA | AK061965.1 | - | 7 - 8 | 559 | 108 | |
| AK066930.1 | - | 20 - 21 | 2774 | 108 | ||
More Information
Set Membership: |
|

LOC4341008
Genomic Map