Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788862.2 | XP_015644348.1 | 21 - 22 | 2951 | 103 | |
| XM_015788863.2 | XP_015644349.1 | 21 - 22 | 2927 | 103 | ||
| XM_015788864.2 | XP_015644350.1 | 20 - 21 | 2846 | 103 | ||
| GenBank mRNA | AK061965.1 | - | 8 - 9 | 711 | 101 | |
| AK066930.1 | - | 21 - 22 | 2928 | 103 | ||
More Information
Set Membership: |
|

LOC4341008
Genomic Map