Assay Details
| Gene Symbol: LOC4341008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788863.2 | XP_015644349.1 | 4 - 5 | 1210 | 73 | |
| GenBank mRNA | AK066930.1 | - | 4 - 5 | 1211 | 73 | |
More Information
Set Membership: |
|

LOC4341008
Genomic Map