Assay Details
| Gene Symbol: LOC4341072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788603.2 | XP_015644089.1 | 10 - 11 | 1125 | 117 | |
| XM_015788604.2 | XP_015644090.1 | 11 - 12 | 1055 | 117 | ||
| XM_015788605.2 | XP_015644091.1 | 9 - 10 | 929 | 117 | ||
| GenBank mRNA | AK064330.1 | - | 10 - 11 | 1116 | 117 | |
| AK121622.1 | - | 10 - 11 | 3165 | 117 | ||
More Information
Set Membership: |
|

LOC4341072
Genomic Map