Assay Details
| Gene Symbol: LOC4341072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788603.2 | XP_015644089.1 | 5 - 6 | 732 | 117 | |
| XM_015788604.2 | XP_015644090.1 | 6 - 7 | 662 | 117 | ||
| XM_015788605.2 | XP_015644091.1 | 6 - 7 | 662 | 117 | ||
| GenBank mRNA | AK064330.1 | - | 5 - 6 | 723 | 117 | |
More Information
Set Membership: |
|

LOC4341072
Genomic Map