Assay Details
| Gene Symbol: LOC4341072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788603.2 | XP_015644089.1 | 7 - 8 | 832 | 148 | |
| XM_015788604.2 | XP_015644090.1 | 8 - 9 | 762 | 148 | ||
| GenBank mRNA | AK064330.1 | - | 7 - 8 | 823 | 148 | |
| AK121622.1 | - | 7 - 8 | 2931 | 148 | ||
More Information
Set Membership: |
|

LOC4341072
Genomic Map