Assay Details
| Gene Symbol: LOC4341072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788603.2 | XP_015644089.1 | 8 - 9 | 929 | 140 | |
| XM_015788604.2 | XP_015644090.1 | 9 - 10 | 859 | 140 | ||
| GenBank mRNA | AK064330.1 | - | 8 - 9 | 920 | 140 | |
More Information
Set Membership: |
|

LOC4341072
Genomic Map