Assay Details
| Gene Symbol: LOC4341305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788492.1 | XP_015643978.1 | 1 - 2 | 278 | 100 | |
| GenBank mRNA | AK068862.1 | - | 1 - 2 | 215 | 100 | |
| AK103977.1 | - | 1 - 2 | 214 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788492.1 transcripts.
Set Membership: |
|

LOC4341305
Genomic Map