Assay Details
| Gene Symbol: LOC4345884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794580.2 | XP_015650066.1 | 6 - 7 | 826 | 78 | |
| GenBank mRNA | AK064030.1 | - | 6 - 7 | 802 | 78 | |
| AK103676.1 | - | 6 - 7 | 813 | 78 | ||
More Information
Set Membership: |
|

LOC4345884
Genomic Map