Assay Details
| Gene Symbol: LOC4346464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756897.2 | XP_015612383.1 | 19 - 20 | 2775 | 81 | |
| GenBank mRNA | AK074022.1 | - | 19 - 20 | 2782 | 81 | |
More Information
Set Membership: |
|

LOC4346464
Genomic Map