Assay Details
| Gene Symbol: LOC4346562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756496.1 | XP_015611982.1 | 1 - 2 | 561 | 96 | |
| XM_015756497.2 | XP_015611983.1 | 1 - 2 | 625 | 96 | ||
| GenBank mRNA | AK065862.1 | - | 1 - 2 | 562 | 96 | |
More Information
Set Membership: |
|

LOC4346562
Genomic Map