Assay Details
| Gene Symbol: LOC4346562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756496.1 | XP_015611982.1 | 3 - 4 | 983 | 64 | |
| XM_015756497.2 | XP_015611983.1 | 3 - 4 | 1047 | 64 | ||
| GenBank mRNA | AK065862.1 | - | 3 - 4 | 984 | 64 | |
More Information
Set Membership: |
|

LOC4346562
Genomic Map