Assay Details
| Gene Symbol: LOC4346661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756498.2 | XP_015611984.1 | 3 - 4 | 544 | 73 | |
| XM_015756499.2 | XP_015611985.1 | 3 - 4 | 544 | 73 | ||
| GenBank mRNA | AK103241.1 | - | 3 - 4 | 531 | 73 | |
More Information
Set Membership: |
|

LOC4346661
Genomic Map