Assay Details
| Gene Symbol: LOC4346661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756498.2 | XP_015611984.1 | 5 - 6 | 643 | 75 | |
| XM_015756499.2 | XP_015611985.1 | 5 - 6 | 643 | 75 | ||
| GenBank mRNA | AK103241.1 | - | 5 - 6 | 630 | 75 | |
More Information
Set Membership: |
|

LOC4346661
Genomic Map