Assay Details
| Gene Symbol: LOC4346897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795802.2 | XP_015651288.1 | 1 - 2 | 222 | 158 | |
| XM_015795803.2 | XP_015651289.1 | 1 - 2 | 222 | 158 | ||
| XM_015795804.2 | XP_015651290.1 | 1 - 2 | 229 | 158 | ||
| GenBank mRNA | AK064341.1 | - | 1 - 2 | 207 | 158 | |
| AK101491.1 | - | 1 - 2 | 148 | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795802.2, XM_015795803.2, XM_015795804.2 transcripts.
Set Membership: |
|

LOC4346897
Genomic Map