Assay Details
| Gene Symbol: LOC4346897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795802.2 | XP_015651288.1 | 5 - 6 | 826 | 125 | |
| XM_015795804.2 | XP_015651290.1 | 5 - 6 | 833 | 125 | ||
| GenBank mRNA | AK064341.1 | - | 6 - 7 | 777 | 125 | |
| AK101491.1 | - | 5 - 6 | 752 | 125 | ||
More Information
Set Membership: |
|

LOC4346897
Genomic Map