Assay Details
| Gene Symbol: LOC4346897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795802.2 | XP_015651288.1 | 6 - 7 | 871 | 116 | |
| XM_015795804.2 | XP_015651290.1 | 6 - 7 | 878 | 116 | ||
| GenBank mRNA | AK064341.1 | - | 7 - 8 | 822 | 116 | |
| AK101491.1 | - | 6 - 7 | 797 | 116 | ||
More Information
Set Membership: |
|

LOC4346897
Genomic Map