Assay Details
| Gene Symbol: LOC4346897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795802.2 | XP_015651288.1 | 8 - 9 | 1210 | 127 | |
| XM_015795803.2 | XP_015651289.1 | 7 - 8 | 1177 | 127 | ||
| XM_015795804.2 | XP_015651290.1 | 8 - 9 | 1226 | 127 | ||
| GenBank mRNA | AK064341.1 | - | 9 - 10 | 1170 | 127 | |
| AK101491.1 | - | 8 - 9 | 1145 | 127 | ||
More Information
Set Membership: |
|

LOC4346897
Genomic Map