Assay Details
| Gene Symbol: LOC4347635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757135.2 | XP_015612621.1 | 2 - 3 | 1000 | 60 | |
| GenBank mRNA | AB101649.1 | - | 1 - 2 | 55 | 60 | |
More Information
Set Membership: |
|

LOC4347635
Genomic Map