Assay Details
Gene Symbol: LOC4347697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756538.2 | XP_015612024.1 | 10 - 11 | 2122 | 105 | |
GenBank mRNA | AK103074.1 | - | 10 - 11 | 2097 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
