Assay Details
Gene Symbol: LOC4347697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756538.2 | XP_015612024.1 | 5 - 6 | 877 | 64 | |
XM_015756539.2 | XP_015612025.1 | 5 - 6 | 878 | 64 | ||
GenBank mRNA | AK103074.1 | - | 5 - 6 | 852 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
