Assay Details
| Gene Symbol: LOC4347697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756538.2 | XP_015612024.1 | 9 - 10 | 2002 | 104 | |
| XM_015756539.2 | XP_015612025.1 | 9 - 10 | 2003 | 104 | ||
| GenBank mRNA | AK103074.1 | - | 9 - 10 | 1977 | 104 | |
More Information
Set Membership: |
|

LOC4347697
Genomic Map