Assay Details
| Gene Symbol: LOC4347954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757715.2 | XP_015613201.1 | 12 - 13 | 1978 | 86 | |
| GenBank mRNA | AK112062.1 | - | 2 - 3 | 402 | 86 | |
More Information
Set Membership: |
|

LOC4347954
Genomic Map