Assay Details
| Gene Symbol: LOC4347954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757715.2 | XP_015613201.1 | 17 - 18 | 2432 | 73 | |
| GenBank mRNA | AK112062.1 | - | 7 - 8 | 856 | 73 | |
More Information
Set Membership: |
|

LOC4347954
Genomic Map