Assay Details
| Gene Symbol: LOC4348739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758634.2 | XP_015614120.1 | 13 - 14 | 1774 | 93 | |
| GenBank mRNA | AK121209.1 | - | 13 - 14 | 1781 | 93 | |
More Information
Set Membership: |
|

LOC4348739
Genomic Map