Assay Details
| Gene Symbol: LOC4348739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758634.2 | XP_015614120.1 | 1 - 2 | 484 | 109 | |
| GenBank mRNA | AK121209.1 | - | 1 - 2 | 491 | 109 | |
More Information
Set Membership: |
|

LOC4348739
Genomic Map