Assay Details
Gene Symbol: LOC4348805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759021.2 | XP_015614507.1 | 4 - 5 | 739 | 119 | |
XM_015759022.2 | XP_015614508.1 | 4 - 5 | 752 | 119 | ||
XM_015759023.1 | XP_015614509.1 | 4 - 5 | 735 | 119 | ||
XM_015759024.2 | XP_015614510.1 | 4 - 5 | 742 | 119 | ||
GenBank mRNA | AK068079.1 | - | 4 - 5 | 736 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
