Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 13 - 14 | 1249 | 105 | |
| XM_015758827.1 | XP_015614313.1 | 9 - 10 | 714 | 105 | ||
| GenBank mRNA | AK318583.1 | - | 13 - 14 | 1348 | 105 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map