Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 19 - 20 | 2145 | 66 | |
| XM_015758827.1 | XP_015614313.1 | 15 - 16 | 1610 | 66 | ||
| GenBank mRNA | AK318583.1 | - | 19 - 20 | 2244 | 66 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map