Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 20 - 21 | 2221 | 110 | |
| XM_015758827.1 | XP_015614313.1 | 16 - 17 | 1686 | 110 | ||
| GenBank mRNA | AK318583.1 | - | 20 - 21 | 2320 | 110 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map