Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 25 - 26 | 2875 | 95 | |
| XM_015758827.1 | XP_015614313.1 | 21 - 22 | 2340 | 95 | ||
| GenBank mRNA | AK318583.1 | - | 24 - 25 | 3597 | 95 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map