Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 26 - 27 | 3113 | 80 | |
| XM_015758827.1 | XP_015614313.1 | 22 - 23 | 2578 | 80 | ||
| GenBank mRNA | AK318583.1 | - | 25 - 26 | 3835 | 80 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map