Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 4 - 5 | 477 | 61 | |
| GenBank mRNA | AK318583.1 | - | 4 - 5 | 576 | 61 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map