Assay Details
| Gene Symbol: LOC4349051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758970.2 | XP_015614456.1 | 4 - 5 | 820 | 96 | |
| XM_015758972.2 | XP_015614458.1 | 4 - 5 | 845 | 96 | ||
| GenBank mRNA | AK070828.1 | - | 4 - 5 | 733 | 96 | |
| AK101024.1 | - | 4 - 5 | 821 | 96 | ||
More Information
Set Membership: |
|

LOC4349051
Genomic Map