Assay Details
| Gene Symbol: LOC4349881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762132.2 | XP_015617618.1 | 1 - 2 | 288 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4349881| Gene Symbol: LOC4349881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762132.2 | XP_015617618.1 | 1 - 2 | 288 | 71 | |
Set Membership: |
|