Assay Details
| Gene Symbol: LOC4350004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761555.2 | XP_015617041.1 | 4 - 5 | 578 | 109 | |
| GenBank mRNA | AK073816.1 | - | 4 - 5 | 581 | 109 | |
More Information
Set Membership: |
|

LOC4350004
Genomic Map