Assay Details
Gene Symbol: LOC4330328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768461.2 | XP_015623947.1 | 1 - 2 | 410 | 87 | |
GenBank mRNA | AK069103.1 | - | 1 - 2 | 318 | 87 | |
FJ666318.1 | - | 1 - 2 | 317 | 87 | ||
U78782.1 | - | 1 - 2 | 218 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
