Assay Details
| Gene Symbol: LOC4330328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768461.2 | XP_015623947.1 | 6 - 7 | 727 | 70 | |
| GenBank mRNA | AK069103.1 | - | 6 - 7 | 635 | 70 | |
| FJ666318.1 | - | 6 - 7 | 634 | 70 | ||
| U78782.1 | - | 6 - 7 | 535 | 70 | ||
More Information
Set Membership: |
|

LOC4330328
Genomic Map