Assay Details
| Gene Symbol: LOC9266378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770235.2 | XP_015625721.1 | 4 - 5 | 486 | 66 | |
| GenBank mRNA | AK106197.1 | - | 4 - 5 | 472 | 66 | |
More Information
Set Membership: |
|

LOC9266378
Genomic Map