Assay Details
| Gene Symbol: LOC4330426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771852.2 | XP_015627338.1 | 10 - 11 | 2286 | 130 | |
| GenBank mRNA | AK072471.1 | - | 10 - 11 | 2287 | 130 | |
More Information
Set Membership: |
|

LOC4330426
Genomic Map