Assay Details
| Gene Symbol: LOC4330426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771852.2 | XP_015627338.1 | 13 - 14 | 2740 | 73 | |
| GenBank mRNA | AK063335.1 | - | 3 - 4 | 396 | 73 | |
| AK072471.1 | - | 13 - 14 | 2742 | 73 | ||
More Information
Set Membership: |
|

LOC4330426
Genomic Map