Assay Details
| Gene Symbol: LOC4330787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771634.1 | XP_015627120.1 | 6 - 7 | 1144 | 68 | |
| GenBank mRNA | AK064259.1 | - | 6 - 7 | 1128 | 68 | |
| AK068256.1 | - | 6 - 7 | 1126 | 68 | ||
More Information
Set Membership: |
|

LOC4330787
Genomic Map