Assay Details
| Gene Symbol: LOC4331167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769899.2 | XP_015625385.1 | 1 | 894 | 81 | |
| GenBank mRNA | AK059979.1 | - | 1 | 845 | 81 | |
More Information
Set Membership: |
|

LOC4331167
Genomic Map